2-Bromo-5-nitrotoluene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-2-methyl-4-nitrobenzene

M-Clarity™ quality level = MQ100

247103-25G 247103-5G
SIAL247103-25GEA 79.9 GBP
SIAL247103-25G SIAL247103-5G
2-Bromo-5-nitrotoluene, Sigma-Aldrich®
2-Bromo-5-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 140…143 °C (17 mmHg)
Melting Pt: 78 °C
MDL Number: MFCD00007281
CAS Number: 7149-70-4
EINECS: 230-481-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR