2,3,6-Trifluorobenzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: α-Bromo-2,3,6-trifluorotoluene, 2-(Bromomethyl)-1,3,4-trifluorobenzene

007504-1G 007504-5G 007504-10G
FLUO007504-1GEA 0 GBP
FLUO007504-1G FLUO007504-5G FLUO007504-10G
2,3,6-Trifluorobenzyl bromide
2,3,6-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00061208
CAS Number: 151412-02-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR