3-Bromo-p-toluic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
264660-25G
264660-5G
SIAL264660-25GEA
94.2
GBP
SIAL264660-25G
SIAL264660-5G
3-Bromo-p-toluic acid, Sigma-Aldrich®
3-Bromo-p-toluic acid
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol |
MDL Number:
MFCD00002488 CAS Number: 7697-26-9 EINECS: 231-712-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...