4'-Bromoacetophenone
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
002964-100G
002964-500G
FLUO002964-100GEA
0
GBP
FLUO002964-100G
FLUO002964-500G
4'-Bromoacetophenone
4'-Bromoacetophenone
Formula:
C₈H₇BrO MW: 199.05 g/mol Boiling Pt: 255 °C (1013 hPa) Melting Pt: 49…52 °C Density: 1.736 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000105 CAS Number: 99-90-1 EINECS: 202-799-3 UN: 2811 ADR: 6.1,II Merck Index: 13,01387 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...