α-Bromo-2-chloro-5-fluorotoluene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

556319-10G 556319-1G
SIAL556319-10GEA 355 GBP
SIAL556319-10G SIAL556319-1G
α-Bromo-2-chloro-5-fluorotoluene, Sigma-Aldrich®
α-Bromo-2-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
MDL Number: MFCD01631419
CAS Number: 81778-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR