α-Bromo-2-chloro-3,6-difluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005600-1G 005600-5G
FLUO005600-1GEA 0 GBP
FLUO005600-1G FLUO005600-5G
α-Bromo-2-chloro-3,6-difluorotoluene
α-Bromo-2-chloro-3,6-difluorotoluene
Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 50 °C
MDL Number: MFCD01631446
CAS Number: 90292-67-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR