1-Bromo-4-butylbenzene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Bromophenyl)butane

B2561-A.10 B2561-A.50
XXXXB2561-A.10EA 0 GBP
XXXXB2561-A.10 XXXXB2561-A.50
1-Bromo-4-butylbenzene
1-Bromo-4-butylbenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: –26 °C
Density: 1,2347 g/cm³ (20 °C)
MDL Number: MFCD00040934
CAS Number: 41492-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR