2,5-Dimethoxybenzenesulphonyl chloride

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dimethoxybenzene-1-sulphonyl chloride

D4166.10 D4166.50
XXXXD4166.10EA 0 GBP
XXXXD4166.10 XXXXD4166.50
2,5-Dimethoxybenzenesulphonyl chloride
2,5-Dimethoxybenzenesulphonyl chloride
Formula: C₈H₉ClO₄S
MW: 236.68 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00024872
CAS Number: 1483-28-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR