3-Methoxythiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Methoxybenzenethiol, 1-Methoxy-3-mercaptobenzene, 3-Mercaptoanisol, 3-Methoxybenzene-1-thiol, 3-Methoxyphenyl mercaptan, m-Methoxythiophenol

001309-25G 001309-100G
FLUO001309-25GEA 0 GBP
FLUO001309-25G FLUO001309-100G
3-Methoxythiophenol
3-Methoxythiophenol
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 223 °C (1013 hPa)
Density: 1.136 g/cm³ (20 °C)
MDL Number: MFCD00004841
CAS Number: 15570-12-4
EINECS: 239-617-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR