Diphenylamine ≥98.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: N-PhenylbenzeneamineN-Phenylaniline

23543.182EA 99.9 GBP
23543.182
Diphenylamine ≥98.0%, AnalaR NORMAPUR® analytical reagent
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 53…54 °C
Density: 1.165 g/cm³ (20 °C)
Flash Pt: 153 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
Merck Index: 13,03349
https://uk.vwr.com/stibo/web/std.lang.all/76/36/2297636.jpg

Order Now

Specification Test Results

Assay Min. 98.0 %
Melting point 52.5 - 54.0 °C
Aniline Max. 0.1 %
Ignition residue (SO4) Max. 0.03 %
Insolubility in ethanol 96 % vol Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR