4-Aminophenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Hydroxyaniline
M-Clarity™ quality level = MQ200
SIALA71328-5GEA
31.5
GBP
SIALA71328-5G
SIALA71328-500G
SIALA71328-1KG
SIALA71328-100G
4-Aminophenol, Sigma-Aldrich®
4-Aminophenol
Formula:
H₂NC₆H₄OH MW: 109.13 g/mol Boiling Pt: 284 °C (1013 hPa) Melting Pt: ∼186 °C Density: 1.3 g/cm³ (20 °C) Flash Pt: 195 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00007869 CAS Number: 123-30-8 EINECS: 204-616-2 UN: 2512 ADR: 6.1,III Merck Index: 13,00461 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...