4-Aminophenol

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Hydroxyaniline

EHERC10212000EA 66.5 GBP
EHERC10212000
4-Aminophenol
4-Aminophenol
Formula: H₂NC₆H₄OH
MW: 109.13 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: ∼186 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 195 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00007869
CAS Number: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III
Merck Index: 13,00461
https://uk.vwr.com/stibo/web/std.lang.all/04/69/2300469.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR