m-Anisidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-Aminoanisole, 3-Methoxyaniline, 3-Anisidine
M-Clarity™ quality level = MQ200
A88204-100G
A88204-1KG
A88204-250G
A88204-5G
SIALA88204-100GEA
78.2
GBP
SIALA88204-100G
SIALA88204-1KG
SIALA88204-250G
SIALA88204-5G
m-Anisidine, Sigma-Aldrich®
m-Anisidine
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 251 °C (1013 hPa) Melting Pt: –2 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007783 CAS Number: 536-90-3 EINECS: 208-651-4 UN: 2431 ADR: 6.1,III Merck Index: 13,00670 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...