3-(Methylthio)phenyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M3750.5 M3750.25
XXXXM3750.5EA 0 GBP
XXXXM3750.5 XXXXM3750.25
3-(Methylthio)phenyl isothiocyanate
3-(Methylthio)phenyl isothiocyanate
Formula: C₈H₇NS₂
MW: 181.28 g/mol
Boiling Pt: 130 °C (1013 hPa)
Density: 1,22 g/cm³ (20 °C)
MDL Number: MFCD00041079
CAS Number: 51333-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR