3-(Methylthio)phenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009543-1G
009543-5G
FLUO009543-1GEA
0
GBP
FLUO009543-1G
FLUO009543-5G
3-(Methylthio)phenyl isothiocyanate
3-(Methylthio)phenyl isothiocyanate
Formula:
C₈H₇NS₂ MW: 181.28 g/mol Boiling Pt: 130 °C (1013 hPa) Density: 1,22 g/cm³ (20 °C) |
MDL Number:
MFCD00041079 CAS Number: 51333-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...