2-(Methylthio)phenyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M3749.2 M3749.10
XXXXM3749.2EA 0 GBP
XXXXM3749.2 XXXXM3749.10
2-(Methylthio)phenyl isothiocyanate
2-(Methylthio)phenyl isothiocyanate
Formula: C₈H₇NS₂
MW: 181.28 g/mol
Boiling Pt: 270 °C (1013 hPa)
Density: 1,227 g/cm³ (20 °C)
MDL Number: MFCD00041061
CAS Number: 51333-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR