3,5-Dimethoxyphenyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dimethoxy-5-isothiocyanatobenzene

D4793.1 D4793.5
XXXXD4793.1EA 0 GBP
XXXXD4793.1 XXXXD4793.5
3,5-Dimethoxyphenyl isothiocyanate
3,5-Dimethoxyphenyl isothiocyanate
Formula: C₉H₉NO₂S
Boiling Pt: 130…132 °C (3 mmHg)
Melting Pt: 51 °C
MDL Number: MFCD00041077
CAS Number: 104968-58-3
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR