3,4-Dimethoxyphenyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dimethoxyphenylisothiocyanate, 1,2-Dimethoxy-4-isothiocyanatobenzene

D4792.2 D4792.10
XXXXD4792.2EA 0 GBP
XXXXD4792.2 XXXXD4792.10
3,4-Dimethoxyphenyl isothiocyanate
3,4-Dimethoxyphenyl isothiocyanate
Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Boiling Pt: 171…172 °C (7 mmHg)
Melting Pt: 48…48.5 °C
MDL Number: MFCD00041075
CAS Number: 33904-04-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR