2,4-Dimethoxyphenyl isocyanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Isocyanato-2,4-dimethoxybenzene

M-Clarity™ quality level = MQ100

251887-1G 251887-5G
SIAL251887-1GEA 66.6 GBP
SIAL251887-1G SIAL251887-5G
2,4-Dimethoxyphenyl isocyanate, Sigma-Aldrich®
2,4-Dimethoxyphenyl isocyanate
Formula: C₉H₉NO₃
MW: 179.18 g/mol
Boiling Pt: 140 °C (11 mmHg)
Melting Pt: 30…31 °C
MDL Number: MFCD00002005
CAS Number: 84370-87-6
EINECS: 282-747-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR