4-Nitrophenyl isocyanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Isocyanato-4-nitrobenzene

M-Clarity™ quality level = MQ100

269425-25G 269425-5G
SIAL269425-25GEA 169 GBP
SIAL269425-25G SIAL269425-5G
4-Nitrophenyl isocyanate, Sigma-Aldrich®
4-Nitrophenyl isocyanate
Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 55…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007306
CAS Number: 100-28-7
EINECS: 202-836-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR