3-Nitrophenyl isocyanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Isocyanato-3-nitrobenzene, 3-Nitrophenylisocyanate

M-Clarity™ quality level = MQ100

269417-25G 269417-5G
SIAL269417-25GEA 280 GBP
SIAL269417-25G SIAL269417-5G
3-Nitrophenyl isocyanate, Sigma-Aldrich®
3-Nitrophenyl isocyanate
Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
Boiling Pt: 120…125 °C (0.7 mmHg)
Melting Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007220
CAS Number: 3320-87-4
EINECS: 222-025-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR