Ethyl-4-iodobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

E2213.10 E2213.50
XXXXE2213.10EA 0 GBP
XXXXE2213.10 XXXXE2213.50
Ethyl-4-iodobenzoate
Ethyl 4-iodobenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 281 °C (1013 hPa)
Density: 1,655 g/cm³ (20 °C)
MDL Number: MFCD00017344
CAS Number: 51934-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR