Methyl-2-iodobenzoate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2-iodobenzoate, 2-Iodobenzoic acid methyl ester
M-Clarity™ quality level = MQ200
532851-250G
532851-50G
SIAL532851-250GEA
296
GBP
SIAL532851-250G
SIAL532851-50G
Methyl-2-iodobenzoate, Sigma-Aldrich®
Methyl 2-iodobenzoate
Formula:
C₈H₇IO₂ MW: 262.05 g/mol Boiling Pt: 272 °C (1013 hPa) Density: 1.784 g/cm³ (20 °C) |
MDL Number:
MFCD00016351 CAS Number: 610-97-9 EINECS: 210-243-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...