4-Fluorothioanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Fluorophenyl methyl sulphide, 1-Fluoro-4-(methylthio)benzene

003730-25G 003730-100G
FLUO003730-25GEA 0 GBP
FLUO003730-25G FLUO003730-100G
4-Fluorothioanisole
4-Fluorothioanisole
Formula: C₇H₇FS
MW: 142.2 g/mol
Boiling Pt: 184…185 °C (1013 hPa)
Density: 1,167 g/cm³ (20 °C)
MDL Number: MFCD00040829
CAS Number: 371-15-3
EINECS: 206-733-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR