2-Fluorothioanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Fluoro-2-(methylthio)benzene, 2-Fluorophenyl methyl sulphide

019279-5G 019279-25G
FLUO019279-5GEA 0 GBP
FLUO019279-5G FLUO019279-25G
2-Fluorothioanisole
2-Fluorothioanisole
Formula: C₇H₇FS
MW: 142.2 g/mol
MDL Number: MFCD03093762
CAS Number: 655-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR