1,3,5-Trifluoro-2-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Nitro-2,4,6-trifluorobenzene, 2,4,6-Trifluoronitrobenzene

007687-5G 007687-25G 007687-100G
FLUO007687-5GEA 0 GBP
FLUO007687-5G FLUO007687-25G FLUO007687-100G
1,3,5-Trifluoro-2-nitrobenzene
1,3,5-Trifluoro-2-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 172 °C (1013 hPa)
Melting Pt: 3.5 °C
Density: 1.514 g/cm³ (20 °C)
MDL Number: MFCD00014687
CAS Number: 315-14-0
EINECS: 206-248-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR