4-Fluoro-2-nitroaniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

SIAL162558-25GEA 42 GBP
SIAL162558-25G SIAL162558-100G
4-Fluoro-2-nitroaniline, Sigma-Aldrich®
4-Fluoro-2-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007830
CAS Number: 364-78-3
EINECS: 206-666-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR