3,4,5-Trifluoroaniline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
559490-1G
559490-5G
SIAL559490-1GEA
35.8
GBP
SIAL559490-1G
SIAL559490-5G
3,4,5-Trifluoroaniline, Sigma-Aldrich®
3,4,5-Trifluoroaniline
Formula:
C₆H₄F₃N MW: 147.1 g/mol |
MDL Number:
MFCD00083522 CAS Number: 163733-96-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...