2,3,5-Trifluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001823-1G 001823-5G
FLUO001823-1GEA 0 GBP
FLUO001823-1G FLUO001823-5G
2,3,5-Trifluoroaniline
2,3,5-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00083549
CAS Number: 363-80-4
UN: 2941
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR