4-Fluorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Fluoro-4-isothiocyanatobenzene

001283-10G 001283-25G
FLUO001283-10GEA 0 GBP
FLUO001283-10G FLUO001283-25G
4-Fluorophenyl isothiocyanate
4-Fluorophenyl isothiocyanate
Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 26…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004809
CAS Number: 1544-68-9
EINECS: 216-280-4
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR