3,4-Difluorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Difluorophenylisothiocyanate, Isothiocyanic acid 3,4-Difluorophenyl ester

009610-1G 009610-5G
FLUO009610-1GEA 0 GBP
FLUO009610-1G FLUO009610-5G
3,4-Difluorophenyl isothiocyanate
3,4-Difluorophenyl isothiocyanate
Formula: C₇H₃F₂NS
MW: 171.17 g/mol
MDL Number: MFCD00175109
CAS Number: 113028-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR