4-Fluoro-3-hydroxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009206-1G 009206-5G 009206-25G
FLUO009206-1GEA 0 GBP
FLUO009206-1G FLUO009206-5G FLUO009206-25G
4-Fluoro-3-hydroxybenzoic acid
4-Fluoro-3-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00269641
CAS Number: 51446-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR