3,4-Difluorosalicylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013026-1G 013026-2.5G
FLUO013026-1GEA 0 GBP
FLUO013026-1G FLUO013026-2.5G
3,4-Difluorosalicylic acid
3,4-Difluorosalicylic acid
Formula: C₇H₄F₂O₃
MW: 174.1 g/mol
MDL Number: MFCD03428564
CAS Number: 189283-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR