6-Fluorosalicylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Fluoro-2-hydroxybenzoic acid, 2-Fluoro-6-hydroxybenzoic acid

006236-1G 006236-5G
FLUO006236-1GEA 0 GBP
FLUO006236-1G FLUO006236-5G
6-Fluorosalicylic acid
6-Fluorosalicylic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00052179
CAS Number: 67531-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR