2-Fluoro-5-hydroxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013317-1G 013317-5G
FLUO013317-1GEA 0 GBP
FLUO013317-1G FLUO013317-5G
2-Fluoro-5-hydroxybenzoic acid
2-Fluoro-5-hydroxybenzoic acid
Formula: C₇H₅FO₃
MDL Number: MFCD01862084
CAS Number: 51446-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR