4-(Trifluoromethoxy)benzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

SIAL301396-1GEA 37.2 GBP
SIAL301396-1G
4-(Trifluoromethoxy)benzoic acid, Sigma-Aldrich®
4-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 153.4 °C
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR