3-(Trifluoromethoxy)benzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

301388-1G 301388-5G
SIAL301388-1GEA 52 GBP
SIAL301388-1G SIAL301388-5G
3-(Trifluoromethoxy)benzoic acid, Sigma-Aldrich®
3-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 91…92 °C
MDL Number: MFCD00041501
CAS Number: 1014-81-9
EINECS: 213-802-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR