2,3,5-Trifluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001826-1G 001826-5G 001826-10G
FLUO001826-1GEA 0 GBP
FLUO001826-1G FLUO001826-5G FLUO001826-10G
2,3,5-Trifluorobenzoic acid
2,3,5-Trifluorobenzoic acid
Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
MDL Number: MFCD00061200
CAS Number: 654-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR