2,3,4-Trifluorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
333824-25G
333824-5G
SIAL333824-25GEA
74.2
GBP
SIAL333824-25G
SIAL333824-5G
2,3,4-Trifluorobenzoic acid, Sigma-Aldrich®
2,3,4-Trifluorobenzoic acid
Formula:
C₇H₃F₃O₂ MW: 176.09 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 140…142 °C |
MDL Number:
MFCD00061232 CAS Number: 61079-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...