2-Fluorobenzhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluorobenzohydrazide, 2-Fluorobenzene-1-carbohydrazide, 2-Fluorobenzoic hydrazide, 2-Fluorobenzoic acid hydrazide

006188.10G 006188-25G 006188-5G
FLUO006188.10GEA 48.7 GBP
FLUO006188.10G FLUO006188-25G FLUO006188-5G
2-Fluorobenzhydrazide
2-Fluorobenzhydrazide
Formula: C₇H₇FN₂O
MW: 154.14 g/mol
MDL Number: MFCD00025112
CAS Number: 446-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR