3-Fluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride

001641-25G 001641-100G 001641-250G
FLUO001641-25GEA 0 GBP
FLUO001641-25G FLUO001641-100G FLUO001641-250G
3-Fluorobenzoyl chloride
3-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000670
CAS Number: 1711-07-5
EINECS: 216-977-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR