2,4,5-Trifluorobenzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2,4,5-trifluorobenzoylchloride
001633-5G
001633-25G
FLUO001633-5GEA
0
GBP
FLUO001633-5G
FLUO001633-25G
2,4,5-Trifluorobenzoyl chloride
2,4,5-Trifluorobenzoyl chloride
Formula:
C₇H₂ClF₃O MW: 194.54 g/mol Boiling Pt: 85 °C (17 mmHg) Density: 1.52 g/cm³ (20 °C) |
MDL Number:
MFCD00061204 CAS Number: 88419-56-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...