2,3,4-Trifluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,3,4-trifluorobenzoylchloride

007494-5G 007494-25G 007494-1G
FLUO007494-5GEA 0 GBP
FLUO007494-5G FLUO007494-25G FLUO007494-1G
2,3,4-Trifluorobenzoyl chloride
2,3,4-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Boiling Pt: 143…144 °C (130 mmHg)
Density: 1.522 g/cm³ (20 °C)
MDL Number: MFCD00075078
CAS Number: 157373-08-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR