2,3,4,5-Tetrafluorobenzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2,3,4,5-tetrafluorobenzoylchloride
007335-5G
007335-25G
FLUO007335-5GEA
0
GBP
FLUO007335-5G
FLUO007335-25G
2,3,4,5-Tetrafluorobenzoyl chloride
2,3,4,5-Tetrafluorobenzoyl chloride
Formula:
C₇HClF₄O MW: 212.53 g/mol Boiling Pt: 53…54 °C (1013 hPa) Density: 1.58 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00075164 CAS Number: 94695-48-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...