2,4,5-Trifluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001871-10G 001871-50G
FLUO001871-10GEA 0 GBP
FLUO001871-10G FLUO001871-50G
2,4,5-Trifluorobenzaldehyde
2,4,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 174 °C (1013 hPa)
Density: 1.408 g/cm³ (20 °C)
MDL Number: MFCD00061196
CAS Number: 165047-24-5
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR