2,3,6-Trifluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007477-1G 007477-5G 007477-25G
FLUO007477-1GEA 0 GBP
FLUO007477-1G FLUO007477-5G FLUO007477-25G
2,3,6-Trifluorobenzaldehyde
2,3,6-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00061195
CAS Number: 104451-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR