2,3,5-Trifluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
007954-5G
007954-10G
FLUO007954-5GEA
0
GBP
FLUO007954-5G
FLUO007954-10G
2,3,5-Trifluorobenzaldehyde
2,3,5-Trifluorobenzaldehyde
Formula:
C₇H₃F₃O MW: 160.1 g/mol Boiling Pt: 165 °C (1013 hPa) Density: 1.407 g/cm³ (20 °C) |
MDL Number:
MFCD00083538 CAS Number: 126202-23-1 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...