3-(Trifluoromethoxy)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-(Trifluoromethoxy)phenyl)metaboric acid, (3-(Trifluoromethoxy)phenyl)boronic acid, 3-(Trifluoromethoxy)benzeneboronic acid, Dihydroxy(3-(trifluoromethoxy)phenyl)borane

013042-25G 013042-5G
FLUO013042-25GEA 0 GBP
FLUO013042-25G FLUO013042-5G
3-(Trifluoromethoxy)phenylboronic acid
3-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
MDL Number: MFCD01320697
CAS Number: 179113-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR