2,4-Difluorophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

011029-5G 011029-25G
FLUO011029-5GEA 0 GBP
FLUO011029-5G FLUO011029-25G
2,4-Difluorophenylboronic acid
2,4-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01318998
CAS Number: 144025-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR