Thiomersal

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt

EMR231010 EMR231025 EMR231100
EURLEMR231010EA 0 GBP
EURLEMR231010 EURLEMR231025 EURLEMR231100
Thiomersal
Thiomersal
Formula: 2-(C₂H₅HgS)C₆H₄CO₂Na
MW: 404.82 g/mol
Melting Pt: 232…233 °C
Flash Pt: 250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00013062
CAS Number: 54-64-8
EINECS: 200-210-4
UN: 2025
ADR: 6.1,II
Merck Index: 12,09451
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621fb9ecf6f0ee8129acf49c938a6167645828da&vid=a4d9481c20c7c69ffe3a642fbe6cae8c3dfb117f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR